Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Flavivirus Capsid Protein Antibody, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313372
                        
                            
                        
                        
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
Flavivirus Capsid Protein Polyclonal antibody specifically detects Flavivirus Capsid Protein in Virus samples. It is validated for Western BlotSpecifications
| Flavivirus Capsid Protein | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Virus | |
| Purified | 
| Western Blot | |
| Unconjugated | |
| C protein, Capsid protein C, Core protein, Flavivirus group Capsid | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Flavivirus Capsid Protein. The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            