Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavivirus group antigen Antibody (D1-4G2-4-15 (4G2)) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP25270902 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25270902 0.2 mg
NBP25270925 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25270902 Supplier Novus Biologicals Supplier No. NBP2527090.2MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Flavivirus group antigen Monoclonal specifically detects Flavivirus group antigen in Monkey, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Functional, Immunofluorescence, Binding Activity, CyTOF-ready.

Specifications

Antigen Flavivirus group antigen
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Classification Monoclonal
Clone D1-4G2-4-15 (4G2)
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry-Paraffin, Immunofluorescence 1:10 - 1:500, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias DEN 1, DEN1, Dengue hemorrhagic fever, Dengue type 1 virus, DENV 1, DENV1
Host Species Mouse
Immunogen Dengue Virus type 2 antigens.
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognises flavivirus group specific antigens (Dengue virus, West Nile Virus, Japanese Encephalitis, Zika virus etc). It binds to the fusion loop at the extremity of domain II of protein E.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.