Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flavivirus group antigen Antibody (D1-4G2-4-15 (4G2)), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252709MFV610
Description
Flavivirus group antigen Monoclonal antibody specifically detects Flavivirus group antigen in Virus samples. It is validated for Flow CytometrySpecifications
Flavivirus group antigen | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Flow Cytometry | |
D1-4G2-4-15 (4G2) | |
Flow Cytometry | |
Envelope Protein, Protein E | |
This recombinant Flavivirus group antigen Antibody (D1-4G2-4-15 (4G2)) was prepared from Dengue Virus type 2 antigens. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction