Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavivirus Group Antigen Mouse anti-Virus, Clone: D1-4G2-4-15, MilliporeSigma™

Catalog No. MAB10216I25 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10216I25 25 μg
MAB10216I10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB10216I25 Supplier MilliporeSigma Supplier No. MAB10216I25UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Flavivirus Group Antigen Monoclonal antibody specifically detects Flavivirus Group Antigen in Virus samples. It is validated for Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flavivirus Group Antigen
Applications Immunofluorescence
Classification Monoclonal
Clone D1-4G2-4-15
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Dengue virus type 2 antigens.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.