Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Flavopiridol hydrochloride
GREENER_CHOICE

Catalog No. 309410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-941-0 10 mg
30-945-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-941-0 Supplier Tocris Bioscience™ Supplier No. 3094/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cdk inhibitor; potently inhibits cdk2 and cdk9

Cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 6 - 25 nM and 84 - 200 nM for cdk9 and cdk2, respectively, and <0.85 μM for cdk1, cdk4, cdk5 and cdk7). Induces cell cycle arrest at G1 and G2 phase. Potently inhibits the growth of breast and lung cancer cell lines (IC50 = 25 - 160 nM) in vitro.

Chemical Identifiers

CAS 131740-09-5
Molecular Formula C21H21Cl2NO5

Specifications

Quantity 10 mg
Formula Weight 438.3
Purity >98%
Molecular Weight (g/mol) 438.301
Chemical Name or Material Flavopiridol hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.