Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flexible Collodion, USP, Spectrum™ Chemical
SDP

Catalog No. p-7157284 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-736 100 mL Amber Glass Bottle
18-602-737 500 mL Amber Glass Bottle
18-602-738 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-736 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CO125100ML
Only null left
Add to Cart
Add to Cart

CO125

Spectrum™ Chemical Flexible Collodion, USP is used as a surgical dressing and forms a flexible nitrocellulose membrane. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-70-0 , 64-17-5
Molecular Formula C24H36N8O38
Molecular Weight (g/mol) 1044.57
InChI Key OWWGYSXLINVAES-UHFFFAOYNA-N
IUPAC Name 2,3-bis(nitrooxy)-6-[(nitrooxy)methyl]-5-{[3,4,5-tris(nitrooxy)-6-[(nitrooxy)methyl]oxan-2-yl]oxy}oxan-4-yl nitrate; 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.[O-][N+](=O)OCC1OC(O[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1OC1OC(CO[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1O[N+]([O-])=O

Specifications

CAS Max % 0.1%, 99.9%
Packaging Amber Glass Bottle
Quantity 100 mL
UN Number UN2059
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.