Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont Industrial FlexTech ANSI A4 Cut Resistant Gloves

Catalog No. 17010054
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

FlexTech ANSI A4 Cut Resistant Gloves with a foam nitrile palm and ESD properties to protect equipment and processes.

  • Anti-static properties that rapidly neutralize the electric discharge for the worker and ESD properties that protect product equipment and workpieces
  • The ultra-lightweight liner provides excellent dexterity as an 18 gauge thin glove with an ANSI Cut Level A4 protection to reduce hand injuries and increased abrasion resistance and durability for prolonged use
  • Foam nitrile palm for grip and touchscreen compatibility
  • Offers ANSI 4 puncture and abrasion resistance

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance EN 16350:2014, EN 1149-2:1997
Coated Area Palm
Applications Industrial
Coating Material Nitrile Foam
Color Gray
Cut Resistance Rating ANSI A4
Disposable Reusable
Exterior Finish Fully Textured
Material Nylon, HPPE, Fiberglass, Lycra, Stainless Steel
Product Type Cut Resistant Glove
Size X-Large
For Use With (Application) Metal Stamping, Electronics Assembly, Small Parts Handling and Assembly
Model Y9258
Resistant To Cut Resistant
Yarn Gauge 18
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.