Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Flextril™ 211 Nitrile Chemical Gloves

Catalog No. 19185770 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-185-770 Medium
19-185-769 Small
19-185-771 Large
19-185-772 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-185-770 Supplier Honeywell™ Supplier No. 333150E/8M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ergonomically designed gloves with a nitrile barrier provide both chemical and ANSI Level A1 cut protection.

  • Superior chemical and cut protection
  • Secure wet and dry grip
  • Enhanced dexterity and comfort
  • Sweat management features
  • Cost-effective
  • Increased durability
  • Compliant with food contact

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance FDA 21 CFR 177.2600
Applications Industrial
Color Blue
Exterior Finish Smooth
Latex Free Yes
Material Nitrile
Size Medium
EN388 Standard EN 388:4X21A
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKMOPT
Glove Cuff Style Straight
Grip Design Smooth
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.