Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLG Mouse anti-Human, Unconjugated, Clone: rFLG/1561, Abnova™

Catalog No. 89337834 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-834 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-834 Supplier Abnova Corporation Supplier No. RAB00409
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

FLG Monoclonal antibody specifically detects FLG in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen FLG
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone rFLG/1561
Conjugate Unconjugated
Description filaggrin
Gene FLG
Gene Alias ATOD2
Host Species Mouse
Immunogen Original antibody is raised against recombinant protein corresponding to amino acids 198-288 of human FLG
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2312
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.