Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLI1 Antibody (FLI1/3183), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308652UV
Description
FLI1 Monoclonal antibody specifically detects FLI1 in Human samples. It is validated for ELISA, Protein ArraySpecifications
FLI1 | |
Monoclonal | |
DyLight 350 | |
EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB | |
Recombinant fragment (aa 107-261) of human FLI1 protein with hexa-histidine tag (Uniprot: Q01543 ) | |
100 μg | |
Cancer, Tumor Suppressors | |
2313 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA, Protein Array | |
FLI1/3183 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction