Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLI1 Rabbit anti-Feline, Human, Mouse, Porcine, Rat, Polyclonal, FabGennix

Catalog No. 502207045
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7045 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7045 Supplier Fabgennix Inc Supplier No. FLI101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLI1 Polyclonal antibody specifically detects FLI1 in Feline, Human, Mouse, Porcine, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen FLI1
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Fli1
Gene Accession No. P26323, Q01543
Gene Alias Ewing sarcoma breakpoint region 2; EWSR2; Fli1; Fli-1; Fli-1 proto-oncogene, ETS transcription factor; Friend leukemia integration 1; Friend leukemia integration 1 transcription factor; Friend leukemia virus integration 1; Proto-oncogene Fli-1; Retroviral integration site protein Fli-1; Sic1; SIC-1; transcription factor ERGB
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1-50 on human Friend Leukemia integration 1 transcription factor protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100515914, 101090716, 14247, 2313, 315532
Target Species Feline, Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.