Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ fliC Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5144519

Catalog No. FEPA5144519


Only null left
Add to Cart

Description

Description

fliC Polyclonal Antibody for Western Blot, ELISA

Flagellin is the subunit protein which polymerizes to form the filaments of bacterial flagella.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

fliC
Polyclonal
Unconjugated
flaF; Flagellin; fliC; hag; hase 1-A flagellin; SPA0911
Recombinant Salmonella paratyphi A Flagellin protein (1-495aa).
100 μL
Primary
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot
4 mg/mL
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Rabbit
Protein G
RUO
Bacteria
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.