Learn More
Flightless-1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | Flightless-1 |
---|---|
Applications | Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1926
![]() |
Bethyl Laboratories, Inc
IHC00340T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1925
![]() |
Bethyl Laboratories, Inc
IHC00340 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee
This gene encodes a protein with a gelsolin-like actin binding domain and an N-terminal leucine-rich repeat-protein protein interaction domain. The protein is similar to a Drosophila protein involved in early embryogenesis and the structural organization of indirect flight muscle. The gene is located within the Smith-Magenis syndrome region on chromosome 17.Specifications
Flightless-1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
FLI, FLIL, Fli1, protein flightless-1 homolog | |
FLII | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q13045 | |
2314 | |
Between 425 and 475 | |
Primary | |
4° C | |
FLII |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.