Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLII Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582369
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582369 Supplier Invitrogen™ Supplier No. PA582369
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YILDCWSDVF IWLGRKSPRL VRAAALKLGQ ELCGMLHRPR HATVSRSLEG TEAQVFKAKF KNWDDVLTVD YTRNAEAVLQ SPGLSGKVKR DAEKKDQMKA DLTALFLPRQ PPMSLAEAEQ LMEEWNEDL Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.

Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLII
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FLII
Gene Accession No. Q13045, Q9JJ28
Gene Alias 3632430F08Rik; FLI; Fli1; flightless I actin binding protein; flightless I homolog; flightless I homolog (Drosophila); Flii; FLII actin remodeling protein; FLII, actin remodeling protein; Fliih; FLIL; Protein flightless-1 homolog
Gene Symbols FLII
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FLII. Recombinant protein control fragment (Product #RP-89498).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14248, 2314
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.