Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLII Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521735

Catalog No. PIPA521735


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, H1299, HeLaS3, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (81%), Chicken (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FLII
Polyclonal
Unconjugated
FLII
3632430F08Rik; FLI; Fli1; flightless I actin binding protein; flightless I homolog; flightless I homolog (Drosophila); Flii; FLII actin remodeling protein; FLII, actin remodeling protein; Fliih; FLIL; Protein flightless-1 homolog
Rabbit
Antigen affinity chromatography
RUO
2314
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.88 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q13045
FLII
Recombinant protein encompassing a sequence within the center region of human Flightless I. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.