Learn More
Description
Specifications
Specifications
| Antigen | FLIP |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Alias | CASHFADD-like antiapoptotic molecule 1, CASP8 and FADD-like apoptosis regulator, CASP8AP1, Caspase homolog, Caspase-eight-related protein, Caspase-like apoptosis regulatory protein, caspase-related inducer of apoptosis, CASPER, CasperFLAME1, Cellular FLICE-like inhibitory protein, c-FLIPR, c-FLIPS, CLARPMACH-related inducer of toxicity, FADD-like apoptotic molecule, FLAME, FLAME-1, I-FLICEMRITc-FLIPFLIPc-FLIPL, Inhibitor of FLICE, Usurpin, usurpin beta |
| Gene Symbols | CFLAR |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acids 75-92 (DRKAVETHLLRNPHLVS) of human FLIP was used as immunogen, GenBank no. gi 12643547. This amino acid sequnce is 100% conserved in FLIP long and short, and various FLIP isoforms as denoted in the NCBI BLAST data base. http://www.ncbi.nlm.nih.gov/blast/. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
