Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLIP Monoclonal Antibody (6G11A6)
GREENER_CHOICE

Catalog No. PIMA531680
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531680 Supplier Invitrogen™ Supplier No. MA531680
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31680 has been tested in indirect ELISA.

FLIP (CFLAR) is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells. It acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. Lacks enzymatic (caspase) activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLIP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6G11A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CFLAR
Gene Accession No. O15519
Gene Alias 2310024N18Rik; A430105C05Rik; Cash; CASP8 and FADD like apoptosis regulator; CASP8 and FADD-like apoptosis regulator; CASP8 and FADD-like apoptosis regulator subunit p12; CASP8 and FADD-like apoptosis regulator subunit p43; CASP8AP1; caspase homolog; Caspase-eight-related protein; caspase-like apoptosis regulatory protein; caspase-related inducer of apoptosis; Casper; Cellular FLICE-like inhibitory protein; Cflar; c-Flip; c-FLIPL; c-FLIPR; c-FLIPS; CLARP; ENSMUSG00000072980; FADD-like antiapoptotic molecule 1; FLAME; FLAME1; FLAME-1; Flip; Gm9845; I-FLICE; Inhibitor of FLICE; MACH-related inducer of toxicity; MRIT; testis secretory sperm-binding protein Li 233m; usurpin; usurpin beta
Gene Symbols CFLAR
Host Species Mouse
Immunogen Purified recombinant fragment of human CFLAR (AA: 100-251) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8837
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.