Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLIP, Mouse anti-Human, Clone: NF6, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50 - $706.50
Specifications
Antigen | FLIP |
---|---|
Clone | NF6 |
Concentration | 1 mg/mL |
Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Description
FLIP Monoclonal specifically detects FLIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
FLIP | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Cell Biology, Inhibitors Activators and Regulators, Signal Transduction, Tumor Suppressors | |
PBS containing 10% glycerol | |
8837 | |
Recombinant human FLIP(aa 1-480). | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
NF6 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CASHFADD-like antiapoptotic molecule 1, CASP8 and FADD-like apoptosis regulator, CASP8AP1, Caspase homolog, Caspase-eight-related protein, Caspase-like apoptosis regulatory protein, caspase-related inducer of apoptosis, CASPER, CasperFLAME1, Cellular FLICE-like inhibitory protein, c-FLIPR, c-FLIPS, CLARPMACH-related inducer of toxicity, FADD-like apoptotic molecule, FLAME, FLAME-1, I-FLICEMRITc-FLIPFLIPc-FLIPL, Inhibitor of FLICE, Usurpin, usurpin beta | |
CFLAR | |
IgG1 | |
>95%, by SDS-PAGE | |
Recognizes short (FLIPS) and long (FLIPL) splice variants of human FLIP. This antibody recognizes an epitope in the N-terminal DED region (aa1-194). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title