Description
- Highly inert gold or Siltek™deactivated seals reduce breakdown and adsorption of active compounds, maximizing component transfer to GC column
- Vespel™ ring embedded in top and bottom surface eliminates need for washer
- Very little torque required to make seal, reduces operator variability
Specifications
Specifications
| Type | Gold-Plated |
| Diameter (Metric) Inner | 1.2 mm |
| For Use With (Application) | Ded Design Allows Significantly More Analyses Than Other Seals |
| Packaging Quantity | 10/Pk. |