Learn More
Restek™ Flip Seal™ Dual Vespel™ Ring Inlet Seals
SDP
Catalog No. 06701309 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Gold-Plated
Siltek-Deactivated
Packaging Quantity:
10/Pk.
2/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packaging Quantity
06-701-309 Siltek-Deactivated 10/Pk.
06-701-308 Gold-Plated 10/Pk.
06-701-306 Gold-Plated 2/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-701-309 Supplier Restek™ Supplier No. 23414
Reversible, two-sided design allows significantly more analyses than other seals
  • Highly inert gold or Siltek™deactivated seals reduce breakdown and adsorption of active compounds, maximizing component transfer to GC column
  • Vespel™ ring embedded in top and bottom surface eliminates need for washer
  • Very little torque required to make seal, reduces operator variability
Specifications
Type Siltek-Deactivated
Diameter (Metric) Inner 1.2 mm
For Use With (Application) Ded Design Allows Significantly More Analyses Than Other Seals
Packaging Quantity 10/Pk.