Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FILIP1 Polyclonal Antibody

Catalog No. PIPA571416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571416 Supplier Invitrogen™ Supplier No. PA571416

Rabbit Polyclonal Antibody

FILIP1 Polyclonal Antibody for Western Blot

This gene encodes a filamin A binding protein. The encoded protein promotes the degradation of filamin A and may regulate cortical neuron migration and dendritic spine morphology. Mice lacking a functional copy of this gene exhibit reduced dendritic spine length and altered excitatory signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FILIP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FILIP1
Gene Accession No. Q7Z7B0
Gene Alias filamin A interacting protein 1; filamin-A-interacting protein 1; FILIP; FILIP1; KIAA1275
Gene Symbols FILIP1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human FILIP1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27145
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.