Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ11184/C4orf43 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982680
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2680 Supplier Bioss Supplier No. BS13180R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMA16 is a protein coding gene.

Specifications

Antigen FLJ11184/C4orf43
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TMA16
Gene Accession No. Q96EY4
Gene Alias 1810029B16Rik; C4orf43; LOC100348782; RGD1305222; Tma16; translation machinery associated 16 homolog; translation machinery associated 16 homolog (S. cerevisiae); Translation machinery-associated protein 16; uncharacterized LOC100348782; UPF0534 protein C4orf43; UPF0534 protein C4orf43 homolog
Gene Symbols TMA16
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FLJ11184/C4orf43.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55319
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.