Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLJ11506 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318422
This item is not returnable.
View return policy
Description
FLJ11506 Polyclonal antibody specifically detects FLJ11506 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
FLJ11506 | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
1 mg/ml | |
Western Blot 1:10000 - 1:25000, Immunoprecipitation 50 ug/mg lysate | |
AAGAB, alpha- and gamma-adaptin-binding protein p34, KPPP1, p34, PPKP1A | |
Recognizes region between residues Between Between 265 and 315.. | |
100 μg | |
Signal Transduction | |
79719 | |
Store at 4°C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction