Learn More
FLJ20530/KAONASHI1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300269AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.
This gene encodes a component of prolyl 4-hydroxylase, a key enzyme in collagen synthesis composed of two identical alpha subunits and two beta subunits. The encoded protein is one of several different types of alpha subunits and provides the major part of the catalytic site of the active enzyme. In collagen and related proteins, prolyl 4-hydroxylase catalyzes the formation of 4-hydroxyproline that is essential to the proper three-dimensional folding of newly synthesized procollagen chains. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| FLJ20530/KAONASHI1 | |
| Polyclonal | |
| Unconjugated | |
| INTS8 | |
| C8orf52, chromosome 8 open reading frame 52, FLJ20530, INT8, MGC131633, protein kaonashi-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55656 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q75QN2 | |
| INTS8 | |
| Between 200 and 250 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.