Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLJ21963 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299142B
Description
FLJ21963 Polyclonal specifically detects FLJ21963 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FLJ21963 | |
Polyclonal | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
acyl-CoA synthetase short-chain family member 3, acyl-CoA synthetase short-chain family member 3, mitochondrial, EC 6.2.1, EC 6.2.1.1, FLJ21963 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human FLJ21963. | |
0.1 mL | |
Lipid and Metabolism | |
79611 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction