Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGK494 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100924 Supplier Invitrogen™ Supplier No. PA5100924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SGK494.

FLJ25006 likely exerts a protein serine/threonine kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGK494
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rskr
Gene Accession No. Q96LW2
Gene Alias BC030499; BC037104; cDNA sequence BC030499; Sgk494; sugen kinase 494; uncharacterized serine/threonine-protein kinase SgK494
Gene Symbols Rskr
Host Species Rabbit
Immunogen A synthesized peptide derived from human SGK494(Accession Q96LW2), corresponding to amino acid residues V214-C264.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124923, 216976, 685208
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.