Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLJ33360 Monoclonal Antibody (OTI10B12)
GREENER_CHOICE

Catalog No. PIMA527348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527348 Supplier Invitrogen™ Supplier No. MA527348
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FLJ33360 Monoclonal Antibody for Western Blot, IHC (P)

FLJ33360 is expressed in heart cells and plays a role in the regulation of the transcription of others genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLJ33360
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI10B12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LINC02145
Gene Accession No. 0
Gene Alias FLJ33360; FLJ33360 protein; LINC02145
Gene Symbols LINC02145
Host Species Mouse
Immunogen Full length human recombinant protein of FLJ33360 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401172
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.