Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLJ37770 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563437 Supplier Invitrogen™ Supplier No. PA563437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RYLEQRISIK FCVKLNKSAS ETHHLLKEAY GDEVMSRARV FDWHKRFKEG REDVRDDARS GRPVTHRTDD NIQKVKDLVC S Highest antigen sequence identity to the following orthologs: Mouse - 22%, Rat - 26%.

GVQW3 (GVQW Motif Containing 3) is a Protein Coding gene. An important paralog of this gene is SETMAR. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLJ37770
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LOC100506127
Gene Accession No. Q3ZCU0
Gene Alias GVQW motif-containing protein 3; GVQW3; LOC100506127; Protein GVQW3; putative uncharacterized protein FLJ37770; putative uncharacterized protein FLJ37770-like
Gene Symbols LOC100506127
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FLJ37770.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100506127
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.