Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLJ40125 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309304100UL
Description
FLJ40125 Polyclonal specifically detects FLJ40125 in Human samples. It is validated for Western Blot.Specifications
FLJ40125 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
EC 3.1.3.16, FLJ40125, probable protein phosphatase 1N, protein phosphatase, Mg2+/Mn2+ dependent, 1N (putative) | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human FLJ40125 (NP_001073870.1). Peptide sequence KKKEREKEGREEEEEEEAGRRAPEGPRSLLTAPRRAQRPHGGAEASGGLR | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
147699 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction