Learn More
FLNB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 206851AP150UL

Description
The antibody is specific to FLNB. It can recognize isoforms 1,2,3,6, and 7 of FLNB. It has no cross reaction to FLNA and FLNC.
FOXK1 is expressed in these cells and regulates cell cycle progression through an interaction with its downstream target the cyclin-dependent kinase inhibitor p21 (CIP). Loss of FOXK1 in mice results in growth retardation and a severe impairment in skeletal muscle regeneration following injury. FOXK1 also shows expression in immature tissues of brain, eye, heart, lung and thymus. It also is predomitly expressed in many maligt tissues, such as tumors of the brain, colon and lymph node.Specifications
FLNB | |
Polyclonal | |
Unconjugated | |
FLNB | |
ABP 278, ABP 280 homolog, Actin binding like protein, AOI, Beta filamin, FH1, Filamin 3, Filamin B, Filamin homolog 1, FLN B, FLN1L, FLN3, FLNB, LRS1, SCT, TABP, TAP, Thyroid autoantigen, Truncated ABP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2317, 286940, 306204 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75369, Q80X90 | |
Flnb | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.