Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLNC Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
SDP

Catalog No. 502238713 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-8713 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-8713 Supplier Proteintech Group Inc Supplier No. 284921AP150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Filamin C (FLNC; also known as γ-FLN; ABP-L; FLN2), a muscle-specific filamin and a large actin-cross-linking protein. Human filamins are ∼280 kDa homodimers, each monomer consisting of an N-terminal actin-binding domain followed by 24 immunoglobulin (Ig)-like domains (d1 to d24), the last of which mediates dimerization. FLNC is specifically expressed in cardiomyocytes and skeletal myocytes and is involved in the maintenance of structural integrity. High filamin C associated with better prognosis of prostate cancer, leukemia and breast cancer patients. ( PMID: 25577646, PMID: 30867563, PMID: 31131323)

Specifications

Antigen FLNC
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 300 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 2318
Target Species Human
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.