Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Float Buoys, for 0.1 mL Slide-A-Lyzer™ MINI Dialysis Devices PROMO

Catalog No. PI69588PM
Click to view available options
:
Thermo Scientific™ Float Buoys

Thermo Scientific Slide-A-Lyzer MINI Dialysis Device Float buoys are low density polyethylene (LPDE) rafts designed to securely hold up to 25 Slide-A-Lyzer MINI Dialysis Devices (0.1 mL only) to permit array dialysis.

Float Buoys for Slide-A-Lyzer MINI Dialysis Devices are intended to hold Slide-A-Lyzer MINI Dialysis Devices (0.1 mL size only), whose bottom surfaces are a dialysis interface composed of a cellulose membrane barrier. The float holds the devices so that the bottoms contact the solution in which they are floated for dialysis. LPDE material is not suitable for use at temperatures greater than 50°C and are not designed for use as water bath floats.

Specifications

Content And Storage Store at room temperature.
Membrane Cellulose Membrane, Dialysis
Format MINI Dialysis Devices
Capacity (Metric) 0.1 mL
Product Line Slide-A-Lyzer™
Quantity 4 Buoys
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.