Learn More
Accuform™ Floor Marking Tape
Catalog No. p-7188243 Shop All AccuformNMC Products
Click to view available options
Color:
Black, White
Black, Yellow
Blue, White
Red, White
Dimensions (L x W):
54 ft. x 2 in.
54 ft. x 3 in.
Accuform™ Floor Marking Tape is used to mark specific areas and promote safety by identifying potential hazard areas
  • Vinyl adheres to floors, walls, piping, posts, beams, handrails, stairways, machinery, and equipment
  • Waterproof, and wear-resistant
  • Minor resistance to acids, oils, grease, caustics, alkalis, and most solvents
  • Permanent self-adhesive back with no liner
Specifications
Thickness 8 mil
Dimensions (L x W) 54 ft. x 2 in.
Color Black, White
Material Adhesive Vinyl
Type Marking Tape
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.