Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Florfenicol, 98%

Catalog No. p-7057578
Click to view available options
Quantity:
1 g
5 g

CAS: 73231-34-2 | C12H14Cl2FNO4S | 358.205 g/mol

A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73231-34-2
Molecular Formula C12H14Cl2FNO4S
Molecular Weight (g/mol) 358.205
MDL Number MFCD00864834
InChI Key AYIRNRDRBQJXIF-NXEZZACHSA-N
PubChem CID 114811
ChEBI CHEBI:87185
IUPAC Name 2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O

Specifications

Type Florfenicol
Physical Form Powder
Melting Point 151°C to 156°C
Color White
Quantity 1 g
Flash Point >110°C (230°F)
Boiling Point 618°C
Merck Index 14,4109
Solubility Information Soluble in ethanol to 25mM and in DMSO to 100mM
Formula Weight 358.21
Percent Purity 98%
Chemical Name or Material Florfenicol
Show More Show Less
RTECSNumber DB6034000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.