Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Florfenicol, 98%

Click to view available options
Quantity:
1g
5g
73231-34-2
C12H14Cl2FNO4S
358.205
MFCD00864834
AYIRNRDRBQJXIF-NXEZZACHSA-N
114811
CHEBI:87185
2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O
73231-34-2
C12H14Cl2FNO4S
358.205
MFCD00864834
AYIRNRDRBQJXIF-NXEZZACHSA-N
114811
CHEBI:87185
2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O
  • A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis
Merck Index 14,4109
Solubility Information Soluble in ethanol to 25mM and in DMSO to 100mM
Formula Weight 358.21
Physical Form Powder
Percent Purity 98%
Boiling Point 618°C
Color White
Melting Point 152°C to 156°C
Quantity 1g
Chemical Name or Material Florfenicol

RTECSNumber : DB6034000