Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Florisil™, 100-200 mesh

Click to view available options
Quantity:
250g
1000g
1343-88-0
MgO3Si
100.39
MFCD00078249
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Magnesium silicate
1343-88-0
MgO3Si
100.39
MFCD00078249
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Magnesium silicate
Applications
Florisil is employed for polyether adsorbent, which has applications of adsorption and decoloring, adsorbent in gas chromatography, in vitamin analysis, antibiotic processing, anti caking agent, used in cosmetics as talcum & baby powder, general modified plastics, mid-range insulation materials, EVA shoes material.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents, hydrogen fluoride.
Quantity 250g
Mesh Size 100 to 200 mesh
Chemical Name or Material Florisil™