Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Florisil™, 60 to 100 Mesh, Grade A, Reagents

Catalog No. 01337240
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-240 3 kg
01-337-239 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-240 Supplier Reagents Holdings Llc Supplier No. C22083003KG
Only null left
Add to Cart
Add to Cart

Florisil™, 60 to 100 Mesh, Grade A, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1343-88-0

Specifications

Grade Reagent
Packaging Poly Pail
Quantity 3 kg
Material Magnesia Silica
Molecular Formula MgO3Si
Physical Form Solid
Packing Material Poly Bottle
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N
IUPAC Name silicon(4+) magnesium(2+) trioxidandiide
Molecular Weight (g/mol) 100.39
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.