Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flotillin-1 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179022IR
Description
Flotillin-1 Polyclonal specifically detects Flotillin-1 in Human, Mouse samples. It is validated for Western Blot.Specifications
Flotillin-1 | |
Polyclonal | |
Western Blot | |
flotillin 1, flotillin-1, integral membrane component of caveolae | |
Rabbit | |
Affinity Purified | |
RUO | |
10211 | |
Store at 4C in the dark. |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FLOT1 | |
The immunogen recognized by this antibody maps to a region between residue 377 and 427 of human Flotillin 1 using the numbering given in entry NP_005794.1 (GeneID 10211). | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction