Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flotillin-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$336.16 - $618.75
Specifications
| Antigen | Flotillin-1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Flotillin-1 Polyclonal specifically detects Flotillin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Flotillin-1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 10211 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HQRAIMAHMTVEEIYKDRQKFSEQVFKVASSDLVNMGISVVSYTLKDIHDDQDYLHSLGKARTAQVQKDARIGEAEAKRDAGIREAKAKQEKVSAQYLSEIEMAKAQRDYELKKAAYDIEVNTRRAQADLAYQLQVAKTKQQIEEQR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| flotillin 1, flotillin-1, integral membrane component of caveolae | |
| FLOT1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human Flotillin-1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title