Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Flotillin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596534 Supplier Invitrogen™ Supplier No. PA596534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HDDQDYLHSL GKARTAQVQK DARIGEAEAK RDAGIREAKA KQEKVSAQYL SEIEMAKAQR DYELKKAAYD IEVNTRRAQA DLAYQLQVAK TKQQIEEQRV QVQVVERAQQ VAVQEQEIAR REKELEARVR KPAEAERYKL ERLAEAEKSQ LIMQAEAEAA SVRMRGEAEA FAIGARARAE AEQMAKKAEA FQLYQEAAQL DMLLEKLPQV AEEISGPLTS ANKITLVSSG SGTMGAAKVT GEVLDILTRL PESVERLTGV SISQVNHKPL RTA Positive Samples: HepG2, HeLa, 293T, A-549, Mouse lung, Mouse heart, Mouse brain, Rat heart, Rat brain; Cellular Location: Cell membrane, Endosome, Melanosome, Membrane, Peripheral membrane protein, caveola.

Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flotillin 1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FLOT1
Gene Accession No. O08917, O75955, Q9Z1E1
Gene Alias DADB-118P11.2; Flot1; flotillin 1; Flotillin-1; I79_013152; integral membrane component of caveolae; OTTHUMP00000164855; Rareg; Reg2; REG-2; reggie-2
Gene Symbols FLOT1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10211, 14251, 64665
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.