Learn More
Invitrogen™ Flotillin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA519713
Description
For Western Blot, this antibody has non-specific bands at 35 kDa. Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with many species based on sequence homology. Store antibody at 4ºC for 1-2 weeks. For long-term storage, store at -20ºC.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
Specifications
Flotillin 1 | |
Polyclonal | |
Unconjugated | |
FLOT1 | |
DADB-118P11.2; Flot1; flotillin 1; Flotillin-1; I79_013152; integral membrane component of caveolae; OTTHUMP00000164855; Rareg; Reg2; REG-2; reggie-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10211, 14251 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
O08917, O75955 | |
FLOT1 | |
Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human Flotillin 1. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.