Learn More
Flotillin-1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303422AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Pig, Orangutan, Monkey and Chimpanzee
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.Specifications
Flotillin-1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O08917, O75955 | |
FLOT1 | |
Between 377 and 427 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
FLOT1 | |
OTTHUMP00000164855; flotillin-1; integral membrane component of caveolae | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10211, 14251 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.