Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flotillin-1 Rabbit anti-Human, Mouse, Clone: S08-0G5, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | Flotillin-1 |
---|---|
Clone | S08-0G5 |
Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Flotillin-1 Monoclonal antibody specifically detects Flotillin-1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Flotillin-1 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
flotillin 1, flotillin-1, integral membrane component of caveolae | |
A synthetic peptide of human Flotillin-1 (Uniprot#: O75955) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S08-0G5 | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Endocrinology, Signal Transduction | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
10211 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title