Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flotillin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Flotillin 1 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0321
![]() ![]() |
Proteintech Group Inc
155711AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes an protein that localizes to the caveolae, which are small domains on the inner cell membranes. This protein plays a role in vesicle trafficking and cell morphology. Alternative splicing results in multiple transcript variants.Specifications
Flotillin 1 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O08917, O75955, Q9Z1E1 | |
10211, 14251, 64665 | |
Flotillin 1 Fusion Protein Ag7902 | |
Primary | |
-20°C | |
FLOT1 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FLOT1, flotillin 1 | |
Flot1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title