Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Flotillin 1 Recombinant Rabbit Monoclonal Antibody (JB19-45)
GREENER_CHOICE

Catalog No. PIMA534713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534713 Supplier Invitrogen™ Supplier No. MA534713
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: PC-3M, rat brain tissue, human lung cancer tissue, human liver tissue, human spleen tissue, HUVEC.

Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flotillin 1
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Recombinant Monoclonal
Clone JB19-45
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene FLOT1
Gene Accession No. O75955
Gene Alias DADB-118P11.2; Flot1; flotillin 1; Flotillin-1; I79_013152; integral membrane component of caveolae; OTTHUMP00000164855; Rareg; Reg2; REG-2; reggie-2
Gene Symbols FLOT1
Host Species Rabbit
Immunogen Synthetic peptide within Human Flotillin 1 aa 378-427.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10211
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.