Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Flotillin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517178
Description
It is not recommended to aliquot this antibody.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling.
Specifications
| Flotillin 2 | |
| Polyclonal | |
| Unconjugated | |
| FLOT2 | |
| AI573412; ECS1; ECS-1; epidermal surface antigen; ESA; ESA1; FLOT2; flotillin 2; Flotillin 2 (epidermal surface antigen 1); flotillin-2; M17S1; membrane component chromosome 17 surface marker 1; Membrane component chromosome 17 surface marker 1 homolog; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1); OTTHUMP00000163513; Reg1; REG-1; Reggie-1; reggie1-1; reggie-2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14252, 2319, 83764 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 24 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q14254, Q60634, Q9Z2S9 | |
| FLOT2 | |
| Synthetic peptide corresponding residue surrounding Leu294 of human flotillin-2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction