Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Flotillin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115198 Supplier Invitrogen™ Supplier No. PA5115198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ESA.

Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flotillin 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene FLOT2
Gene Accession No. Q14254, Q60634, Q9Z2S9
Gene Alias AI573412; ECS1; ECS-1; epidermal surface antigen; ESA; ESA1; FLOT2; flotillin 2; Flotillin 2 (epidermal surface antigen 1); flotillin-2; M17S1; membrane component chromosome 17 surface marker 1; Membrane component chromosome 17 surface marker 1 homolog; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1); OTTHUMP00000163513; Reg1; REG-1; Reggie-1; reggie1-1; reggie-2
Gene Symbols FLOT2
Host Species Rabbit
Immunogen A synthesized peptide derived from human FLOT2(Accession Q14254), corresponding to amino acid residues K160-T210.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14252, 2319, 83764
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.