Learn More
MilliporeSigma™ Supelco™ Flow Controller for Flash-Chromatography Assembly with Threaded Joints
Catalog No. p-7191249 Shop All MilliporeSigma Supelco Products
Click to view available options
Joint Size:
ST/NS 29/32
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Flash Chromatography
Joint Size ST/NS 29/32
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.