Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ FlowTACS Flow Cytometry Apoptosis Detection Kit
GSA_VASDP

Catalog No. 481760K Shop All Novus Products
Change view
Click to view available options
Quantity:
60 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
481760K 60 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 481760K Supplier R&D Systems™ Supplier No. 481760K
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Streptavidin-Fluorescein and all the agents necessary for generating positive controls

Supplied with a streptavidin-FITC conjugate for detection of DNA fragmentation providing a fluorescent readout

Can be used for the detection of apoptosis using flow cytometry, direct visualization using a fluorescent microscope or for quantitation using a fluorometer

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 60 Tests
Content And Storage The product requires storage at both -20 to -70° C and 2 - 8° C. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.