Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FLRG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560983
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560983 Supplier Invitrogen™ Supplier No. PA560983
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGSGNPAPGG VCWLQQGQEA TCSLVLQTDV TRAECCASGN IDTAWSNLTH PGNKINLLGF LGLVHCLPCK DSCDG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

Follistatin-like 3 (FSTL3) is a gene located on chromosome 19p13.3 and encodes a secreted glycoprotein that belongs to the follistatin-related protein family. FSTL3 functions as an antagonist to members of the transforming growth factor-beta (TGF-β) superfamily, particularly inhibiting activin and myostatin signaling. This inhibitory action is crucial for regulating muscle growth, fertility, and inflammation. FSTL3 is expressed in various tissues, including the placenta, gonads, and skeletal muscle, where it modulates physiological processes by binding to and neutralizing the activities of its target ligands. Dysregulation of FSTL3 has been implicated in conditions such as muscle wasting, metabolic syndrome, and reproductive disorders, reflecting its role in maintaining tissue homeostasis and function. Research into FSTL3 focuses on its potential therapeutic applications in diseases involving TGF-β pathway dysregulation, offering insights into strategies for modulating growth factor activities in clinical settings.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FLRG
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FSTL3
Gene Accession No. O95633
Gene Alias E030038F23Rik; FLRG; FLRGFSRP; follistatin like 3; follistatin-like 3; follistatin-like 3 (secreted glycoprotein); follistatin-like protein 3; Follistatin-related gene protein; follistatin-related protein 3; FSRP; FSTL3; UNQ674/PRO1308
Gene Symbols FSTL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FLRG. Recombinant protein control fragment (Product #RP-100061).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10272
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.